Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046226.1 | 4 | 1435 | |||
NR_046227.1 | 3 | 1145 | ||||
GenBank mRNA | AK095119.1 | - | 3 | 1145 | ||
AK124928.1 | - | 4 | 1435 |