Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUB-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120537.1 | 4 | 1936 | |||
NR_120538.1 | 3 | 1879 | ||||
GenBank mRNA | AK055772.1 | - | 4 | 1935 |