Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110827.1 | 1, 2 | 502 | |||
GenBank mRNA | BC034818.1 | - | 1, 2 | 502 |