Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023921.2 | 2 | 1028 | |||
NR_023922.2 | 2 | 769 | ||||
NR_023923.2 | 3 | 737 | ||||
NR_023924.2 | 2 | 828 | ||||
GenBank mRNA | BC080582.1 | - | 2 | 755 | ||
BX247982.1 | - | 2 | 498 | |||
BX447332.2 | - | 2 | 952 | |||
BX647964.1 | - | 3 | 707 | |||
HQ845197.1 | - | 3 | 723 | |||
HQ845198.1 | - | 3 | 723 | |||
HQ845199.1 | - | 2 | 808 | |||
JQ979294.1 | - | 3 | 723 | |||
KJ545484.1 | - | 2 | 664 | |||
KJ574270.1 | - | 2 | 455 | |||
KJ574271.1 | - | 2 | 588 | |||
KJ574272.1 | - | 2 | 587 | |||
KJ574273.1 | - | 2 | 747 | |||
KJ574274.1 | - | 2 | 791 | |||
KJ599455.1 | - | 2 | 521 | |||
KJ599456.1 | - | 2 | 778 | |||
KJ599457.1 | - | 2 | 480 | |||
KJ599458.1 | - | 1 | 274 | |||
KJ599459.1 | - | 3 | 696 | |||
KJ599460.1 | - | 2 | 822 | |||
KJ599461.1 | - | 3 | 706 |