Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNG-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104124.1 | 5 | 514 | |||
NR_104125.1 | 4 | 396 | ||||
GenBank mRNA | AK124066.1 | - | 4 | 396 | ||
BG756823.1 | - | 5 | 503 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA982190.1 | - | 4 | 397 | Chr. 12 - 67989445 - 68020108 [+] on Build GRCh38 |