Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00928 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027074.1 | 1 | 403 | |||
NR_027075.1 | 1 | 403 | ||||
NR_027076.1 | 1 | 403 | ||||
NR_027077.1 | 1 | 403 | ||||
GenBank mRNA | AK058110.1 | - | 1 | 385 | ||
BC039350.1 | - | 1 | 403 | |||
BC102002.1 | - | 1 | 122 | |||
BC102003.1 | - | 1 | 122 | |||
BC102004.1 | - | 1 | 122 | |||
BC102005.1 | - | 1 | 122 | |||
BC113965.1 | - | 1 | 122 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX096939.1 | - | 1 | 387 | Chr. 15 - 89510068 - 89524018 [-] on Build GRCh38 |