Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037883.1 | 3, 4 | 466 | |||
NR_131233.1 | 3, 4 | 347 | ||||
NR_131234.1 | 3, 4 | 391 | ||||
NR_131235.1 | 3, 4 | 438 | ||||
GenBank mRNA | AI879178.1 | - | 3, 4 | 445 | ||
AK096498.1 | - | 3, 4 | 448 | |||
AL036798.3 | - | 3, 4 | 380 | |||
BF700938.1 | - | 3, 4 | 464 | |||
CK004193.1 | - | 3, 4 | 348 |