Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037883.1 | 4 | 649 | |||
NR_131233.1 | 4 | 530 | ||||
NR_131234.1 | 4 | 574 | ||||
NR_131235.1 | 4 | 621 | ||||
GenBank mRNA | AK096498.1 | - | 4 | 631 | ||
CK004193.1 | - | 4 | 531 |