Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC55338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130914.1 | 1 | 113 | |||
GenBank mRNA | AK002104.1 | - | 1 | 702 | ||
AK023417.1 | - | 1 | 114 | |||
AK023539.1 | - | 1 | 1059 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU134573.1 | - | 1 | 114 | Chr. 5 - 87217892 - 87239201 [-] on Build GRCh38 |