Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LL22NC03-75H12.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109967.1 | 2 | 442 | |||
GenBank mRNA | BC037972.1 | - | 2 | 442 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI823165.1 | - | 2 | 442 | Chr. 22 - 47461492 - 47487111 [-] on Build GRCh38 |