Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120646.1 | 5 | 1264 | |||
GenBank mRNA | BC014575.1 | - | 5 | 1268 |