Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF25-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103750.1 | 2 | 301 | |||
GenBank mRNA | AB016899.1 | - | 1 | 605 | ||
AB016900.1 | - | 2 | 301 | |||
BC112215.1 | - | 1 | 36 | |||
BC143766.1 | - | 1 | 36 | 9052280 |