Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033938.1 | 1 | 401 | |||
GenBank mRNA | BC035396.1 | - | 1 | 392 | ||
BX161461.1 | - | 1 | 401 |