Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRM2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027274.1 | 5 | 1118 | |||
NR_027275.1 | 5 | 592 | ||||
GenBank mRNA | AK056063.1 | - | 3 | 1524 | ||
AK098626.1 | - | 5 | 1118 | |||
AK125806.1 | - | 1 | 3285 | |||
DB448439.2 | - | 5 | 592 |