Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003367.3 | 2 | 504 | |||
GenBank mRNA | BC041065.1 | - | 1 | 263 | ||
BX280136.1 | - | 2 | 483 | |||
M34428.1 | - | 2 | 477 |