Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110086.1 | 5 | 602 | |||
NR_110087.1 | 3 | 224 | ||||
NR_110088.1 | 2 | 179 | ||||
GenBank mRNA | BG213706.1 | - | 1 | 100 | ||
BM152701.1 | - | 3 | 219 | |||
CB217567.1 | - | 2 | 179 |