Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130777.1 | 7 | 1072 | |||
GenBank mRNA | AK098570.1 | - | 7 | 1089 |