Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024256.1 | 1 | 836 | |||
NR_104327.1 | 2 | 414 | ||||
NR_104336.1 | 2 | 324 | ||||
GenBank mRNA | AK128810.1 | - | 1 | 835 | ||
BC036297.1 | - | 2 | 414 |