Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024090.2 | 8 | 935 | |||
NR_109786.1 | 8 | 1046 | ||||
NR_109787.1 | 7 | 811 | ||||
NR_109788.1 | 7 | 872 | ||||
GenBank mRNA | AK094858.1 | - | 7 | 760 | ||
BC042856.1 | - | 8 | 890 | |||
BC044727.1 | - | 8 | 890 | |||
BX537735.1 | - | 7 | 872 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | EB388790.1 | - | 6 | 681 | Chr. 21 - 20743587 - 20803027 [-] on Build GRCh38 |