Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP3-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103443.1 | 3 | 242 | |||
GenBank mRNA | AK302139.1 | - | 3 | 242 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DC397927.1 | - | 3 | 242 | Chr. 3 - 9216895 - 9218087 [+] on Build GRCh38 |