Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120578.1 | 3 | 1467 | |||
GenBank mRNA | AK095187.1 | - | 3 | 1468 |