Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111950.1 | 2 | 989 | |||
NR_111952.1 | 2 | 989 | ||||
GenBank mRNA | AB007962.1 | - | 2 | 657 |
Gene Symbol: LINC00623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024510.2 | 2 | 680 | |||
GenBank mRNA | AK311378.1 | - | 2 | 670 | ||
BC094809.1 | - | 2 | 602 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM019493.1 | - | 1 | 526 | Chr. 1 - 16888936 - 16889644 [-] on Build GRCh38 |