Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120366.1 | 4 | 488 | |||
GenBank mRNA | AL357200.1 | - | 1 | 350 |