Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109888.1 | 7 | 2467 | |||
GenBank mRNA | BC047601.1 | - | 7 | 2467 |