Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832663.1 | - | 16 | 2927 |
Gene Symbol: LINC01449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110832.1 | 1 | 98 | |||
GenBank mRNA | BC033981.1 | - | 1 | 104 |