Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024281.1 | 1 | 2221 | |||
GenBank mRNA | AF052108.1 | - | 1 | 641 | ||
AK055056.1 | - | 1 | 1137 | |||
AK091593.1 | - | 1 | 1970 | |||
AL832535.1 | - | 1 | 946 | |||
BC042097.1 | - | 1 | 92 |