Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110687.1 | 2 | 356 | |||
NR_110688.1 | 2 | 312 | ||||
NR_110689.1 | 2 | 271 | ||||
GenBank mRNA | BI259890.1 | - | 2 | 244 | ||
BX426127.2 | - | 2 | 208 | |||
CN415214.1 | - | 2 | 255 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF038869.1 | - | 2 | 255 | Chr. 19 - 27793483 - 27798282 [+] on Build GRCh38 |