Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_110533.1 | 6 | 1080 | |||
GenBank mRNA | X91348.1 | - | LIT1227 DGCR5 |
6 | 1053 |