Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX2-1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103710.1 | 2 | 821 | |||
GenBank mRNA | BX161496.1 | - | 2 | 783 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX367709.2 | - | 2 | 779 | Chr. 14 - 36519316 - 36522225 [+] on Build GRCh38 |