Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC37BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125981.1 | - | 1 | 155 |
Gene Symbol: LOC107133515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015341.2 | 1 | 154 | |||
GenBank mRNA | DB024747.1 | - | 1 | 154 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR740624.1 | - | 1 | 120 | Chr. 17 - 30576499 - 30601284 [+] on Build GRCh38 |