Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPEAR-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026547.1 | 3 | 813 | |||
NR_026548.1 | 2 | 739 | ||||
GenBank mRNA | AF426269.1 | - | 3 | 813 | ||
AF426270.1 | - | 2 | 739 | |||
BC016690.2 | - | 2 | 337 | |||
BC064517.1 | - | 2 | 337 | |||
DA908807.1 | - | 2 | 334 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE384098.1 | - | 2 | 316 | Chr. 21 - 44517637 - 44518860 [+] on Build GRCh38 |