Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126414.1 | 2 | 166 | |||
GenBank mRNA | CB960713.1 | - | 193 | |||
CB961243.1 | - | 193 | ||||
DA128020.1 | - | 2 | 178 |