Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR133A1HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110369.1 | 2 | 162 | |||
GenBank mRNA | AF086463.1 | - | 2 | 299 | ||
BX648566.1 | - | 2 | 163 |