Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120475.1 | 3 | 671 | |||
GenBank mRNA | BC030099.2 | - | 3 | 673 |