Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM44-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047573.1 | 1 | 59 | |||
NR_047574.1 | 1 | 59 | ||||
NR_047575.1 | 1 | 59 | ||||
GenBank mRNA | BI088778.1 | - | 1 | 59 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK124168.1 | - | 1 | 324 | Chr. 3 - 194584003 - 194958854 [+] on Build GRCh38 |