Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105378349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134313.1 | 1 | 95 | |||
NR_134314.1 | 1 | 95 | ||||
GenBank mRNA | DB069022.1 | - | 1 | 95 |