Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM163A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006711174.2 | XP_006711237.1 | 1, 2 | 259 | ||
XM_006711175.2 | XP_006711238.1 | 1, 2 | 249 | |||
XM_006711176.2 | XP_006711239.1 | 1, 2 | 259 | |||
XM_011509217.1 | XP_011507519.1 | 1, 2 | 240 | |||
XM_017000386.1 | XP_016855875.1 | 1, 2 | 259 | |||
XM_017000387.1 | XP_016855876.1 | 1, 2 | 259 | |||
XM_017000388.1 | XP_016855877.1 | 1, 2 | 259 | |||
XM_017000389.1 | XP_016855878.1 | 1, 2 | 259 | |||
XM_017000390.1 | XP_016855879.1 | 1, 2 | 259 | |||
XM_017000391.1 | XP_016855880.1 | 1, 2 | 259 | |||
GenBank mRNA | AK302665.1 | - | 1, 2 | 249 | ||
BC033359.1 | - | 1, 2 | 209 |