Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100134040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL162052.1 | - | 1 | 419 |
Gene Symbol: LOC101929998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127966.1 | - | 1 | 838 |