Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038985.1 | 1 | 225 | |||
GenBank mRNA | AK091031.1 | - | 1 | 102 | ||
AK294518.1 | - | 1 | 110 |