Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCBP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040047.1 | 2 | 525 | |||
NR_040048.1 | 2 | 272 | ||||
NR_040049.1 | 2 | 272 | ||||
GenBank mRNA | AK055993.1 | - | 2 | 194 | ||
BC034773.1 | - | 2 | 205 | |||
DA274215.1 | - | 2 | 447 |