Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFDN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027906.1 | 2 | 1960 | |||
GenBank mRNA | AB016898.1 | - | 2 | 1873 | ||
AK091690.1 | - | 3 | 1862 |