Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104147.1 | 1 | 489 | |||
GenBank mRNA | AK097794.1 | - | 1 | 489 |