Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015404.1 | 2 | 1701 | |||
GenBank mRNA | AF229831.1 | - | 2 | 206 | ||
AK096935.1 | - | 2 | 1701 | |||
AK124158.1 | - | 1 | 1571 | |||
BC007973.2 | - | 2 | 348 | |||
BC015633.2 | - | 2 | 717 | |||
BC023547.2 | - | 2 | 1352 | |||
BC136540.1 | - | 2 | 1620 | |||
JQ794500.1 | - | 2 | 1804 |