Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF4A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109949.1 | 2, 3 | 349 | |||
GenBank mRNA | AF143870.1 | - | 2, 3 | 360 |