Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125422.1 | 2 | 69 | |||
GenBank mRNA | BC015456.1 | - | 2 | 72 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG179358.1 | - | 70 | Chr. - - [-] on Build |