Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135279.1 | 1 | 42 | |||
GenBank mRNA | AF426268.1 | - | 1 | 30 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA394976.1 | - | 1 | 42 | Chr. 21 - 45234340 - 45257267 [+] on Build GRCh38 |