Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117100.1 | 5 | 1119 | |||
GenBank mRNA | DQ515897.1 | - | 5 | 1119 | ||
DQ515898.1 | - | 1 | 489 | |||
DQ515899.1 | - | 1 | 489 | |||
JX003864.1 | - | 1 | 390 | |||
JX003866.1 | - | 1 | 390 |