Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs.457407-USP11 fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092258.1 | - | 597 |
Gene Symbol: NR2F1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109818.1 | 2, 3 | 300 | |||
NR_109819.1 | 1, 2 | 862 | ||||
NR_109822.1 | 1, 2 | 862 | ||||
NR_109825.1 | 2, 3 | 967 | ||||
GenBank mRNA | AK095719.1 | - | 2, 3 | 278 | ||
AK124699.1 | - | 1, 2 | 912 | |||
AK296281.1 | - | 1, 2 | 549 | |||
AK311323.1 | - | 1, 2 | 575 | |||
BC044619.1 | - | 2, 3 | 443 | |||
BC048999.1 | - | 2, 3 | 443 | |||
BC144330.1 | - | 1, 2 | 1462 | |||
BC144331.1 | - | 1, 2 | 1462 |