Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110773.1 | 2 | 320 | |||
NR_110774.1 | 2 | 184 | ||||
GenBank mRNA | BX108181.1 | - | 2 | 184 | ||
HY048010.1 | - | 2 | 321 |