Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01053 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120427.1 | 2 | 278 | |||
GenBank mRNA | AB593160.1 | - | 2 | 278 | ||
BU176203.1 | - | 2 | 255 |